Learn More
DiscoverX PathHunter™ CHO-K1 PPYR1 (NPY4R) β-Arrestin Cell Line
The PathHunter™ CHO-K1 PPYR1 (NPY4R) β-Arrestin Cell Line measures PPYR1 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0233C2
Encompass
Product Code. 17352789
  • 33718.50€ / 2 ampoules
Estimated Shipment: 22-01-2025
Description
The PathHunter™ PPYR1 (NPY4R) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. pancreatic polypeptide) based activation of PPYR1 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged PPYR1 and EA-tagged β-Arrestin 2. Activation of PPYR1 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
β-Arrestin recruitment
Cell Lines
2 Vials
AY268432
CHO-K1
Pancreatic polypeptide
PPYR1 (NPY4R)
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.