Learn More
DiscoverX PathHunter™ DLD1 IL7R/CRLF2 Dimerization Cell Line
The PathHunter™ DLD1 IL7R/CRLF2 Dimerization Cell Line measures ligand-induced dimerization of IL7R/CRLF2 . This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-1019C13
Encompass
Product Code. 17304249
  • 33718.50€ / 2 ampoules
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The PathHunter™ IL7R/CRLF2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TSLP) induced dimerization of IL7R/CRLF2 receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside CRLF2 target protein fused to EA. Upon ligand-induced dimerization of IL7R/CRLF2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Dimerization
Cell Lines
2 Vials
NM_002185.3
DLD1
TSLP
IL7R/CRLF2