Learn More
DiscoverX PathHunter™ HEK 293 GPR152 β-Arrestin Orphan GPCR Cell Line
The PathHunter™ HEK 293 GPR152 β-Arrestin Orphan GPCR Cell Line measures GPR152 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0384C1A
Encompass
Product Code. 17363089
  • 33718.50€ / 2 ampoules
Estimated Shipment: 17-01-2025
Description
The PathHunter™ GPR152 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR152 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR152 and EA-tagged β-Arrestin 2. Activation of GPR152 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Orphan receptor with unknown ligand.
Specifications
Specifications
β-Arrestin recruitment
Cell Lines
GPR152
293H
2 Vials
NM_206997.1
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.