Learn More
DiscoverX PathHunter™ HEK 293 IL17RA/IL17RC Dimerization Cell Line
The PathHunter™ HEK 293 IL17RA/IL17RC Dimerization Cell Line measures ligand-induced dimerization of IL17RA/IL17RC . This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 93-0931C1
This item is not returnable.
View return policy
Description
The PathHunter™ IL17RA/IL17RC Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-17a) induced dimerization of IL17RA/IL17RC receptors. This product uses EFC technology. The cell line included overexpresses IL17RC target protein fused to PK alongside IL17RA target protein fused to EA. Upon ligand-induced dimerization of IL17RA/IL17RC , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Dimerization | |
Cell Lines | |
2 Vials | |
NM_014339 |
293H | |
IL17A | |
IL17RA/IL17RC |