Learn More
DiscoverX PathHunter™ HEK 293 IL17RA/IL17RC Dimerization Cell Line
The PathHunter™ HEK 293 IL17RA/IL17RC Dimerization Cell Line measures ligand-induced dimerization of IL17RA/IL17RC . This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0931C1
Encompass
Product Code. 17374029
  • 35218.50€ / 2 ampoules
Estimated Shipment: 07-02-2025
Description
The PathHunter™ IL17RA/IL17RC Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-17a) induced dimerization of IL17RA/IL17RC receptors. This product uses EFC technology. The cell line included overexpresses IL17RC target protein fused to PK alongside IL17RA target protein fused to EA. Upon ligand-induced dimerization of IL17RA/IL17RC , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Dimerization
Cell Lines
2 Vials
NM_014339
293H
IL17A
IL17RA/IL17RC
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.