Learn More
DiscoverX PathHunter™ U2OS BMPR1B/BMPR2 Dimerization Cell Line
The PathHunter™ U2OS BMPR1B/BMPR2 Dimerization Cell Line measures ligand-induced dimerization of BMPR1B/BMPR2 . This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-1053C3
Encompass
Product Code. 17304319
  • 35218.50€ / 2 ampoules
Estimated Shipment: 05-02-2025
Description
The PathHunter™ BMPR1B/BMPR2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. BMP-6) induced dimerization of BMPR1B/BMPR2 receptors. This product uses EFC technology. The cell line included overexpresses BMPR1B target protein fused to PK alongside BMPR2 target protein fused to EA. Upon ligand-induced dimerization of BMPR1B/BMPR2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Dimerization
Cell Lines
2 Vials
NM_001256793.1
U2OS
BMP-6
BMPR1B/BMPR2
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.