Learn More
DiscoverX PathHunter™ U2OS c-Ret-GFRα2 Functional Assay
The PathHunter™ U2OS c-Ret-GFRα2 Functional Assay measures c-Ret-GFRα2 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells. Cyno version.
Brand:  DiscoverX 93-0816C3
Encompass
Product Code. 17333839
  • 35218.50€ / 2 ampoules
Description
The PathHunter™ U2OS c-Ret-GFRα2 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated c-Ret and GFRα2 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged c-Ret and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, GFRα2 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the cyno version of the protein.
Specifications
Specifications
NPR1 Activation
Cell Lines
2 Vials
NM_020975.4
U2OS
Recombinant Human Neurturin
c-Ret-GFRα2
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX PathHunter™ U2OS c-Ret-GFRα2 Functional Assay