Learn More
DiscoverX PathHunter™ U2OS CD86 Ligand Cell Line
The PathHunter™ U2OS CD86 Ligand Cell Line works with Checkpoint signaling cell lines to induce SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 93-1093C3
This item is not returnable.
View return policy
Description
The PathHunter CD86 Ligand cell line is a stable, engineered cell line designed for use with CTLA4 Checkpoint Signaling Cell Lines. When co-cultured, the CD86 expressed in this cell line will induce activity of CTLA4 receptors, which may be assayed through SH2 domain protein recruitment. This product is a companion product designed to work with Immunoglobulin Superfamily Checkpoint Assays. The cell line included overexpresses CD86 cell surface ligand. When co-cultured with the corresponding Checkpoint Signaling Cell Line, the CD86 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Ligand Presenting | |
Cell Lines | |
2 Vials | |
NM_175862.4 |
U2OS | |
Anti-CTLA4 Antibody [14D3] | |
CD86 |