Learn More
DiscoverX PathHunter™ U2OS CD86 Ligand Cell Line
The PathHunter™ U2OS CD86 Ligand Cell Line works with Checkpoint signaling cell lines to induce SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-1093C3
Encompass
Product Code. 17384369
  • 16318.50€ / 2 ampoules
Estimated Shipment: 28-01-2025
Description
The PathHunter CD86 Ligand cell line is a stable, engineered cell line designed for use with CTLA4 Checkpoint Signaling Cell Lines. When co-cultured, the CD86 expressed in this cell line will induce activity of CTLA4 receptors, which may be assayed through SH2 domain protein recruitment. This product is a companion product designed to work with Immunoglobulin Superfamily Checkpoint Assays. The cell line included overexpresses CD86 cell surface ligand. When co-cultured with the corresponding Checkpoint Signaling Cell Line, the CD86 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Ligand Presenting
Cell Lines
2 Vials
NM_175862.4
U2OS
Anti-CTLA4 Antibody [14D3]
CD86
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.