Learn More
DiscoverX PathHunter™ U2OS ErbB4 Functional Assay
The PathHunter™ U2OS ErbB4 Functional Assay measures ErbB4 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 93-0465C3
Description
The PathHunter™ U2OS ErbB4 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated ErbB4 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged ErbB4 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Receptor Activation | |
Cell Lines | |
2 Vials | |
NM_005235 |
U2OS | |
Recombinant Human Neuregulin-1 Beta 2 | |
ErbB4 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.