Learn More
DiscoverX PathHunter™ U2OS GPR22 β-Arrestin Orphan GPCR Cell Line
The PathHunter™ U2OS GPR22 β-Arrestin Orphan GPCR Cell Line measures GPR22 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 93-1085C3
Description
The PathHunter™ GPR22 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR22 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR22 and EA-tagged β-Arrestin 2. Activation of GPR22 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Orphan receptor with unknown ligand.Specifications
β-Arrestin Recruitment | |
Cell Lines | |
GPR22 |
U2OS | |
2 Vials | |
NM_005295.2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.