Learn More
DiscoverX PathHunter™ U2OS GPR85 β-Arrestin Orphan GPCR Cell Line
The PathHunter™ U2OS GPR85 β-Arrestin Orphan GPCR Cell Line measures GPR85 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0409C3
Encompass
Product Code. 17303139
  • 35218.50€ / 2 ampoules
Estimated Shipment: 03-02-2025
Description
The PathHunter™ GPR85 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR85 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR85 and EA-tagged β-Arrestin 2. Activation of GPR85 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Orphan receptor with unknown ligand.
Specifications
Specifications
β-Arrestin recruitment
Cell Lines
GPR85
U2OS
2 Vials
NM_018970
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.