Learn More
DiscoverX PathHunter™ U2OS HTR2C (VGV) β-Arrestin Cell Line
The PathHunter™ U2OS HTR2C (VGV) β-Arrestin Cell Line measures HTR2C (VGV) (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0932C3
Encompass
Product Code. 17304039
  • 35218.50€ / 2 ampoules
Estimated Shipment: 05-02-2025
Description
The PathHunter™ HTR2C (VGV) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR2C GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged HTR2C and EA-tagged β-Arrestin 2. Activation of HTR2C stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
β-Arrestin Recruitment
Cell Lines
2 Vials
NM_000868.2
U2OS
Serotonin/5-HT
HTR2C (VGV)
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.