Learn More
DiscoverX PathHunter™ U2OS HTR2C (VGV) β-Arrestin Cell Line
The PathHunter™ U2OS HTR2C (VGV) β-Arrestin Cell Line measures HTR2C (VGV) (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 93-0932C3
This item is not returnable.
View return policy
Description
The PathHunter™ HTR2C (VGV) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of HTR2C GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged HTR2C and EA-tagged β-Arrestin 2. Activation of HTR2C stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
β-Arrestin Recruitment | |
Cell Lines | |
2 Vials | |
NM_000868.2 |
U2OS | |
Serotonin/5-HT | |
HTR2C (VGV) |