Learn More
DiscoverX PathHunter™ U2OS IL20R1/IL20R2 Dimerization Cell Line
The PathHunter™ U2OS IL20R1/IL20R2 Dimerization Cell Line measures ligand-induced dimerization of IL20R1/IL20R2 . This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-1027C3
Encompass
Product Code. 17324259
  • 33718.50€ / 2 ampoules
Estimated Shipment: 22-01-2025
Description
The PathHunter™ IL20R1/IL20R2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-24) induced dimerization of IL20R1/IL20R2 receptors. This product uses EFC technology. The cell line included overexpresses IL20R2 target protein fused to PK alongside IL20R1 target protein fused to EA. Upon ligand-induced dimerization of IL20R1/IL20R2 , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Dimerization
Cell Lines
2 Vials
NM_014432.3
U2OS
IL-24
IL20R1/IL20R2
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.