Learn More
DiscoverX PathHunter™ U2OS mINSRb Functional Assay
The PathHunter™ U2OS mINSRb Functional Assay measures mINSRb receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells. Mouse version.
Brand:  DiscoverX 93-1017C3
Encompass
Product Code. 17344239
  • 33718.50€ / 2 ampoules
Estimated Shipment: 22-01-2025
Description
The PathHunter™ U2OS mINSRb Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated mINSRb dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged mINSRb and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the mouse version of the protein.
Specifications
Specifications
SH2 Recruitment
Cell Lines
2 Vials
AGH68804.1
U2OS
Insulin, Human
mINSRb
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.