Learn More
DiscoverX PathHunter™ U2OS TrkB Functional Assay
The PathHunter™ U2OS TrkB Functional Assay measures TrkB receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0463C3
Encompass
Product Code. 17353209
  • 33718.50€ / 2 ampoules
Estimated Shipment: 22-01-2025
Description
The PathHunter™ U2OS TrkB Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated TrkB dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged TrkB and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Receptor Activation
Cell Lines
2 Vials
NM_006180
U2OS
Recombinant Human BDNF
TrkB
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.