Learn More
DiscoverX PathHunter™ U2OS TrkB Functional Assay
The PathHunter™ U2OS TrkB Functional Assay measures TrkB receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX 93-0463C3
This item is not returnable.
View return policy
Description
The PathHunter™ U2OS TrkB Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated TrkB dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged TrkB and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Receptor Activation | |
Cell Lines | |
2 Vials | |
NM_006180 |
U2OS | |
Recombinant Human BDNF | |
TrkB |