Learn More
DiscoverX PathHunter™ U2OS TrkC-P75 Functional Assay
The PathHunter™ U2OS TrkC-P75 Functional Assay measures TrkC-P75 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0531C3
Encompass
Product Code. 17373349
  • 35218.50€ / 2 ampoules
Estimated Shipment: 17-03-2025
Description
The PathHunter™ U2OS TrkC-P75 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated TrkC and P75 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged TrkC and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, P75 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Receptor Activation
Cell Lines
2 Vials
NM_001012338
U2OS
Recombinant Human NT-3
TrkC-P75
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
DiscoverX PathHunter™ U2OS TrkC-P75 Functional Assay