Learn More
DiscoverX PathHunter™ U2OS TSHR (Long Isoform) β-Arrestin Cell Line
The PathHunter™ U2OS TSHR (Long Isoform) β-Arrestin Cell Line measures TSHR(L) (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX 93-0760C3
Encompass
Product Code. 17343779
  • 33718.50€ / 2 ampoules
Estimated Shipment: 17-01-2025
Description
The PathHunter™ TSHR(L) β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. TSH) based activation of TSHR GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged TSHR and EA-tagged β-Arrestin 2. Activation of TSHR stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
β-Arrestin Recruitment
Cell Lines
2 Vials
NM_000369.2
U2OS
TSH, Thyroid stimulating hormone, Human
TSHR(L)
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.