Learn More
Invitrogen™ Human PDK-1, His Tag Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ P3001
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
PDK1 (3-phosphoinositide dependent kinase-1) is a serine/threonine protein kinase localized to the plasma membrane. It plays a vital role in insulin-induced glycogen synthesis, PKC activation, and activation of the AGC subfamily of protein kinases. Plays an important role in cellular responses to hypoxia and is important for cell proliferation under hypoxia. Protects cells against apoptosis in response to hypoxia and oxidative stress.
Specifications
Q15118 | |
Kinase Assay | |
5163 | |
Human PDK-1, His Tag | |
5 μg | |
[Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 1, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 1, mitochondrial; B830012B01; D530020C15Rik; isoenzyme 1; kinase isoenzyme 1; lipoamide; mitochondrial pyruvate dehydrogenase, lipoamide, kinase isoenzyme 1; PDH kinase 1; PDHK1; PDK p48; Pdk1; pyruvate dehydrogenase; pyruvate dehydrogenase kinase 1; Pyruvate dehydrogenase kinase isoform 1; pyruvate dehydrogenase kinase, isoenzyme 1; pyruvate dehydrogenase kinase, isozyme 1 | |
PDHK | |
Unconjugated | |
Recombinant | |
Full length | |
Baculovirus | |
Liquid |
See Label | |
50 mM tris with 0.05% Triton X-100, 0.5 mM EDTA, 0.5 mM EGTA, 100 mM NaCl, 20% glycerol, 5 mM DTT and no preservative; pH 7.5 | |
66.9 kDa | |
7.5 | |
For research use only. Not for use in diagnostic procedures | |
PDK1 | |
PDK1 | |
Human | |
His-tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Kinase and inhibitors | |
Serine/threonine kinase |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2075787F | Certificate of Analysis | 21/02/2025 | P3001 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.