Learn More
Afora™ Pear-shaped Schlenk Flask
Add material through a ground glass joint. Afora™ Pear-shaped Schlenk Flask is a reaction vessel typically used in air-sensitive chemistry.
Brand: Afora™ 101/E
This item is not returnable.
View return policy
Description
- Pear-shaped
- Round bottom
- One side-arm with PTFE stopcock key
- Female ground glass joints
Specifications
50 mL | |
PTFE stopcock | |
14/23 | |
50 mm |
Glass | |
Pear | |
Round |