Learn More
Afora™ Pear-shaped Schlenk Flask
Add material through a ground glass joint. Afora™ Pear-shaped Schlenk Flask is a reaction vessel typically used in air-sensitive chemistry.
Brand:  Afora™ 102/E
 View more versions of this product
Product Code. 11976148
  • 157.00€ / Each
Estimated Shipment: 25-03-2025
Description
  • Pear-shaped
  • Round bottom
  • One side-arm with PTFE stopcock key
  • Female ground glass joints
Specifications
Specifications
100 mL
PTFE stopcock
14/23
60 mm
Glass
Pear
Round