Learn More
Pyrex™ Borosilicate Glass Pestle
Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion. Pyrex™ Borosilicate Glass Pestle is ideal for crushing solids into powders for easier handling. Pestles are available in a rough or glazed finish. For use with Pyrex™ Mortars, sold separately.
Brand:  Pyrex 718342R
16.76 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 13471869
  • 19.50€ / Each
Estimated Shipment: 08-08-2025
Description
  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex™ Borosilicate Glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex™ Mortars (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glazed
Borosilicate Glass
110mm
Pestle
Pyrex™ Mortar
Product Title
Pyrex™ Borosilicate Glass Pestle > Length: 110mm; Finish: Glazed