Learn More
Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent
Pierce™ Detergent Compatible Bradford Assay Reagent
Brand: Thermo Scientific™ 23246S
This item is not returnable.
View return policy
Description
The Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent is a quick and ready-to-use modification of the well-known Bradford coomassie dye-binding, colorimetric method for total protein quantitation. Proprietary additives to the Bradford Reagent make it compatible with up to 1% or higher of detergents and lysis reagents that are commonly used in life science research, including Triton™ X-100 and NP-40.Similar to the Bradford method, coomassie dye binds protein in an acidic medium causing an immediate shift in absorption maximum from 465 nm to 595 nm with a concomitant color change from green to blue. In addition, the assay is complete in just 10 minutes.
The protein assay can be performed in either test tube or microplate format. The standard working range is 100-1500ÎĽg/mL with up to 1% detergent (or higher in some cases). Protein concentrations are estimated by reference to absorbances obtained for a series of standard protein dilutions, typically bovine serum albumin (BSA), which are assayed alongside the unknown samples. Because the color response with coomassie dye is non-linear with increasing protein concentration, a standard curve must be completed with each assay. Standards can be used directly without preparing them in the same detergent found in the test samples.
Specifications
Detergent Compatible Bradford Assay Reagent, 15 Test Tube Assays, Protein Quantitation Assay Assay Type, Colorimetric Detection Method, Absorbance, Solution-based Detection Technique, Used For Protein Assays and Analysis | |
Protein assays and analysis | |
4°C | |
15 Assays |
Liquid | |
15 Assay(s) | |
Pierce™ Detergent Compatible Bradford Assay Reagent |
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ Detergent Compatible Bradford Assay Reagent > 15 test tube assays
For Research Use Only. Not for use in diagnostic procedures.