Learn More
Eppendorf™ Pipette Carousel 2
Carries up to six manual single or multi-channel Eppendorf pipettes.
Brand: Eppendorf™ 3116000015
168.99 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Carries up to six manual single or multi-channel Eppendorf pipettes.
- Includes rotatable stand with completely new design that holds all current manual pipettes
- Pipette holders are interchangeable, no tools required
- Robust design that securely holds and protects pipettes
- Handle for easy transport from bench to bench
- When used with optional pipette holder, the carousel can carry the Multipette
Specifications
Manual Single or Multi-channel Eppendorf Pipettes (6) | |
6 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now