Learn More
Lovibond™ Plastic cell
For Comparator 2000+ and CHECKIT comparators
Brand:  Lovibond™ 145505
 View more versions of this product
Product Code. 15663864
  • 23.75€ / Pack of 5
Estimated Shipment: 06-02-2025
Description
For Comparator 2000+ and CHECKIT comparators
Plastic cell, 13.5 mm path length, with grey lid, for Comparator 2000+ and CHECKIT comparators. 
Specifications
Specifications
Set of 5
13.5 mm pathlength
Plastic Cell