Learn More
Lovibond™ Plastic cell
For Comparator 2000+ and CHECKIT comparators
23.75€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15663864
|
Lovibond™
145505 |
Set of 5 |
23.75€
Pack of 5 |
|
|||||
Description
For Comparator 2000+ and CHECKIT comparators
Plastic cell, 13.5 mm path length, with grey lid, for Comparator 2000+ and CHECKIT comparators.
Specifications
13.5 mm pathlength | |
Plastic Cell |