Learn More
KD Scientific Plastic Syringe
Economical and disposable, available in sizes ranging from 1 cc to 50 cc, Luer Slip (LS) or Luer Lock (LL) tip
Brand: KD Scientific 780854
This item is not returnable.
View return policy
Description
- A new line of disposable plastic sterile syringes for all scientific applications
- Norm-ject syringes are the ideal solution for any situation
- Their unique two-part system is latex free and contains no silicone lubricant or rubber
Specifications
Polyethylene (Plunger) | |
Yes | |
Single-use | |
KD Scientific Syringe Pumps | |
Yes |
No Needle Included | |
Syringe | |
20 mL | |
Luer Lock | |
Sterile |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
KD Scientific Plastic Syringe > 20cc luer lock