Learn More
ratiolab™ Multi-Purpose Beakers With Screw-Caps
Use these beakers for a variety of storage and sampling applications. ratiolab™ Multi-Purpose Beakers With Screw-Caps are ideal for sampling, transport and storage of solid, pasty and liquid samples.
Brand:  ratiolab™ 5921183
Product Code. 10691963
  • 94.10€ / Pack of 250
Estimated Shipment: 28-04-2025
Description
Polypropylene (PP) or polystyrene (PS) sample cups with red screw-cap are supplied in non-sterile or sterile and are individually packed.
Specifications
Specifications
Sterile
120 mL
Polypropylene
250 Pk.
Urine Beaker
Product Title
ratiolab™ Multi-Purpose Beakers With Screw-Caps > Capacity: 125mL