Learn More
Cole-Parmer™ Polypropylene Weighing Funnels
Cole-Parmer Polypropylene Weighing Funnels, Description: Size: Small, For Use With: Volumetric flask, Designed to Fit Microwell Plates
Brand: Cole-Parmer™ 06127-02
94.78 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
Cole-Parmer Polypropylene Weighing Funnels, Description: Size: Small, For Use With: Volumetric flask, Designed to Fit Microwell Plates
Specifications
Polypropylene | |
Volumetric flask | |
Weighing Funnel |
Size: Small | |
Designed to Fit Microwell Plates |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now