Learn More
Cole-Parmer™ Essentials Anti-Static Weighing Funnels, Polypropylene
Polypropylene Anti-Static Weighing Funnels ensure complete sample transfer.
Brand: Cole-Parmer™ Essentials 06127-00
127.17 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Autoclavable
- 6 month warranty
- These disposable polypropylene (PP) weighing funnels are designed to fit in volumetric flasks
- One side of the funnel is flattened so you can set the funnel flat on a balance
Specifications
Polypropylene | |
Size: Micro | |
Designed to Fit Microwell Plates |
Autoclavable | |
Volumetric flask | |
Weighing Funnel |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now