Learn More
ratiolab™ Cell-Coun-Tainer Cell Counter Vials
Cell-Coun-Tainer Cell Counter Vials feature a high-precision design made of clear polystyrene. Use these vials with robotic analytical systems.
Brand: ratiolab™ 5515040
This item is not returnable.
View return policy
Description
For Coulter™ and Hycel™ cell counter systems.Specifications
Clear | |
25 mL | |
1000 Pk. |
Polystyrene | |
Lid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
ratiolab™ Cell-Coun-Tainer Cell Counter Vials > Volume: 25mL