Learn More
DiscoverX PrecisION™ hASIC3 Recombinant Stable Cell Line
The PrecisION™ hASIC3 Recombinant Stable Cell Line measures ASIC3 ion channel activity via electrophysiological measurements. This cell line ships as two vials of cryopreserved cells.
Brand:  DiscoverX CYL3055
Encompass
Product Code. 17334919
  • 57743.24€ / 2 ampoules
Estimated Shipment: 22-01-2025
Description
The PrecisION™ hASIC3 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying ASIC3 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses ASIC3 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
Ion Channel Activity
Cell Lines
ASIC3
293H
2 Vials
NM_004769
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.