Learn More
DiscoverX PrecisION™ hKv7.2/hKv7.3 Recombinant Stable Cell Line
The PrecisION™ hKv7.2/hKv7.3 Recombinant Stable Cell Line measures Kv7.2/Kv7.3 ion channel activity via electrophysiological measurements. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX CYL3059
This item is not returnable.
View return policy
Description
The PrecisION™ hKv7.2/hKv7.3 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Kv7.2/Kv7.3 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Kv7.2/Kv7.3 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Ion Channel Activity | |
Cell Lines | |
Kv7.2/Kv7.3 |
CHO-K1 | |
2 Vials | |
Y15065 |