Learn More
DiscoverX PrecisION™ hKv7.3/hKv7.5 Recombinant Stable Cell Line
The PrecisION™ hKv7.3/hKv7.5 Recombinant Stable Cell Line measures Kv7.3/Kv7.5 ion channel activity via electrophysiological measurements. This cell line ships as two vials of cryopreserved cells.
Brand: DiscoverX CYL3060
This item is not returnable.
View return policy
Description
The PrecisION™ hKv7.3/hKv7.5 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying Kv7.3/Kv7.5 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses Kv7.3/Kv7.5 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.Specifications
Ion Channel Activity | |
Cell Lines | |
Kv7.3/Kv7.5 |
CHO-K1 | |
2 Vials | |
NM_004519 |