Learn More
OHAUS™ Probe PT100(A), Stainless Steel (316/304)
GREENER_CHOICE
High Quality Accessories for Hot Plate Stirrers
Brand:  OHAUS™ 30973654
Product Code. 30182893
  • 127.00€ / 1 unit
Only null left
In Stock
Description
  • Accessory for Guardian™ Hotplates and Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For Guardian range
1
Temperature Probe
Probe PT100(A) -40°C to 400°C 25 cm SS 316/304
40°C to 400°C
Stainless Steel (AISI 304)
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Probe PT100(A), Stainless Steel (316/304) > Probe