Learn More
R&D Systems™ Proteome Profiler Human XL Oncology Array
The Proteome Profiler Human XL Oncology Array requires no specialized equipment and eliminates the need for multiple Western blot experiments.
Brand:  R&D Systems™ ARY026
Product Code. 18067631
  • 924.00€ / 1 set
Description
The Proteome Profiler Human XL Oncology Array contains 4 membranes-each spotted in duplicate with 84 different cancer-related antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.
1 Kit
4 Array Membranes, 4-Well Multi-Dish, Array Buffers, Wash Buffer, Detection Antibody Cocktail, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Proteome Profiler Human XL Oncology Array > 1 Kit