missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino Ni-IDA 1000 kit for His-tag Protein Purification in Midi format
Solution for good ratio of His-tag protein yield and purity in Midi format.
43.05€ - 236.00€
Specifications
Bed Volume | 500 μL |
---|---|
Binding Capacity | 5mg |
Binding Property | Batch |
Kit Contents | Buffers |
Matrix | Macroporous Silica |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12778682
|
Macherey-Nagel™
745160.5 |
43.05€
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12788682
|
Macherey-Nagel™
745160.50 |
236.00€
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Protino™ Ni-IDA, the purer alternative for higher yields of His-tag proteins- Chelating group IDA, three protein binding sites
- High protein yield/recovery even from diluted samples
- High protein concentration
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-IDA resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink or swell - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-IDA resin
- Empty 96 well receiver plates available for use with Protino™ Ni-IDA resin
- Ready to use pre-packed 96 well plates filled with dry Protino™ Ni-IDA resin
- Buffers included For purification of polyhistidine-tagged proteins, batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
500 μL | |
Batch | |
Macroporous Silica | |
Ni-TED Protein Purification Kit |
5mg | |
Buffers | |
For Purification of Polyhistidine-Tagged Proteins | |
IDA (iminodiacetic acid) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Protino Ni-IDA 1000 packed columns (5)
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : imidazole, monosodium phosphate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Macherey-Nagel™ Protino Ni-IDA 1000 kit for His-tag Protein Purification in Midi format