Learn More
Quickfit™ PTFE Clamp
For retaining glassware having Quickfit™ conical joints
Brand: Quickfit GC29
35.72 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Used for retaining glassware having Quickfit™ conical joints
- Manufactured from PTFE with encapsulated steel spring to provide firm grip
- Clips are numbered with joint size for easy identification
- Can be used at temperatures up to 250°C
Specifications
Joint Clamp | |
Encapsulated Steel spring | |
29/32 |
PTFE | |
Conical Joints | |
250°C |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Product Title
Quickfit™ PTFE Clamp > For use with: 29/32 conical joint glassware