Learn More
Quickfit™ PTFE Clamp
For retaining glassware having Quickfit™ conical joints
36.75€ - 37.60€
Specifications
Type | Joint Clamp |
---|---|
Material | PTFE |
Includes | Encapsulated Steel spring |
For Use With (Equipment) | Conical Joints |
Temperature (Metric) Resistant | 250°C |
Product Code | Brand | For Use With (Equipment) | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Size | Price | Quantity & Availability | |||||
11941528
|
Quickfit
GC14 |
Conical Joints | 14/23 |
36.75€
Pack of 5 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
11951528
|
Quickfit
GC29 |
Conical Joints | 29/32 |
37.60€
Pack of 5 |
Estimated Shipment: 21-07-2025
Log in to see stock. |
|||||
Description
- Manufactured from PTFE with encapsulated steel spring to provide a firm grip
- Highly resistant to chemical attack
- Can be used at temperatures up to 250°C
- Clips are numbered with joint size for easy identification
Specifications
Joint Clamp | |
Encapsulated Steel spring | |
250°C |
PTFE | |
Conical Joints |
Product Title
Quickfit™ PTFE Clamp