missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human DPPII/QPP/DPP7 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 3438-SE-010
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate, Lys-Pro-AMC (KP-AMC). The specific activity is >4,000 pmol/min/μg, as measured under the described conditions.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
AAH11907 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 64 kDa, reducing conditions; M.W. (theoretical): 54 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
DPP7 | |
Recombinant | |
DPPII/QPP/DPP7 |
Enzyme Activity | |
29952 | |
DPPII/QPP/DPP7 | |
Mouse myeloma cell line, NS0-derived human DPPII/QPP/DPP7 protein Gly22-Leu492, with a C-terminal 10-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human DPPII/QPP/DPP7 Protein, CF > 10ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction