Learn More
R&D Systems™ Proteome Profiler Human Cell Stress Array Kit
The Proteome Profiler Human Cell Stress Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
Brand: R&D Systems™ ARY018
This item is not returnable.
View return policy
Description
The Proteome Profiler Human Cell Stress Array Kit contains 4 membranes - each spotted in duplicate with 26 different cell stress antibodies.
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
1 Kit |
Product Content Correction
Product Title
R&D Systems™ Proteome Profiler Human Cell Stress Array Kit > 1 kit