Learn More
R&D Systems™ Proteome Purify 2 Mouse Serum Protein Immunodepletion Resin
Provides an easy-to-use, high capacity system to reproducibly remove albumin and IgG from mouse serum or plasma
645.00€ - 1140.00€
Specifications
Product Type | Immunodepletion Resin |
---|---|
Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18038101
|
R&D Systems™
MIDR002-040 |
40 Tests |
1140.00€
1 set |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
18078181
|
R&D Systems™
MIDR002-020 |
20 Tests |
645.00€
1 set |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
Description
Analysis of low abundance proteins by even the most sensitive of methods for proteomics greatly benefits from the removal of high abundance proteins that may interfere with their detection.
- Improves visualization of target proteins
- Allows user to increase protein load or concentration
Specifications
Immunodepletion Resin | |
Store the unopened product at 2 - 8° C. Do not use past expiration date. |