Learn More
R&D Systems™ Recombinant Mouse MMP-9 Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ WBC031
This item is not returnable.
View return policy
Description
The Recombinant Mouse MMP-9 Western Blot Standard Protein has been validated for the following applications: Western Blot.Specifications
Western Blot | |
MMP-9 Western Blot Standard | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Unconjugated | |
MMP-9 |
4318 | |
1 Vial | |
Mmp9 | |
Recombinant |
Product Content Correction