missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG Fc Secondary Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74860APC
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Goat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Specifications
IgG Fc | |
Polyclonal | |
PBS | |
Purified Goat IgG Fc fragment | |
0.5 mL | |
Secondary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay (CIA) | |
APC | |
Rabbit | |
Affinity purified | |
RUO | |
Goat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rabbit anti-Goat IgG Fc Secondary Antibody, Allophycocyanin, Novus Biologicals™ > 0.5 mL; APC
Spot an opportunity for improvement?Share a Content Correction