Learn More
R&D Systems™ Rat Luminex™ Discovery Assay
Brand:  R&D Systems™ LXSARM-02
 View more versions of this product
Product Code. 18743677
  • 724.00€ / 1 set
Explore more special offers
Specifications
2 Plex
Rat
Multiplex Cytokine Assay Kits
Store the unopened kit at 2°C to 8°C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Rat Luminex™ Discovery Assay > 2 Plex