missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat Luminex™ Discovery Assay
643.00€ - 2550.00€
Specifications
Species | Rat |
---|---|
Binding Target | Multiplex Cytokine Assay Kits |
Content And Storage | Store the unopened kit at 2°C to 8°C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700018
|
R&D Systems™
LXSARM-08 |
8 Plex |
1955.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711349
|
R&D Systems™
LXSARM-04 |
4 Plex |
1135.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743677
|
R&D Systems™
LXSARM-02 |
2 Plex |
724.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18782747
|
R&D Systems™
LXSARM-11 |
11 Plex |
2550.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18771137
|
R&D Systems™
LXSARM-01 |
1 Plex |
643.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Rat | |
Multiplex Cytokine Assay Kits | |
Store the unopened kit at 2°C to 8°C. Do not use past the expiration date on the label. Avoid freezing microparticles. Protect microparticles from light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Rat Luminex™ Discovery Assay