Learn More
DiscoverX Ready-to-Assay™ 5-HT2A Serotonin Receptor Frozen Cells
The Ready-to-Assay™ 5-HT2A Serotonin Receptor Frozen Cells measure HTR2A (GPCR) activation via an increase in intracellular calcium. Ready-to-Assay kits contain two vials of thaw-and-use cells.
Brand: DiscoverX HTS082RTA
This item is not returnable.
View return policy
Description
The Ready-to-Assay™ 5-HT2A Serotonin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. serotonin/5-HT) based activation of HTR2A GPCR activity via detection of increasing levels of intracellular calcium. The included cells overexpress HTR2A alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of HTR2A , the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the cells in a convenient thaw-and-use format.Specifications
5-HT2A | |
serotonin/5-HT | |
HTR2A |
Frozen Cells | |
2 Vials of Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.