missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cytosine deaminase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Cytosine deaminase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7203-ec-50ug

 View more versions of this product

Product Code. 16027690

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_414871.1
Cytosine deaminase Protein
Research Use Only
codA
Recombinant Protein
E. coli
SNNALQTIIN ARLPGEEGLW QIHLQDGKIS AIDAQSGVMP ITENSLDAEQ GLVIPPFVEP HIHLDTTQTA GQPNWNQSGT LFEGIERWAE RKALLTHDDV KQRAWQTLKW QIANGIQHVR THVDVSDATL TALKAMLEVK QEVAPWIDLQ IVAFPQEGIL SYPNGEALLE EALRLGADVV GAIPHFEFTR EYGVESLHKT FALAQKYDRL IDVHCDEIDD EQSRFVETVA ALAHHEGMGA RVTASHTTAM HSYNGAYTSR LFRLLKMSGI NFVANPLVNI HLQGRFDTYP KRRGITRVKE MLESGINVCF GHDDVFDPWY PLGTANMLQV LHMGLHVCQL MGYGQINDGL NLITHHSART LNLQDYGIAA GNSANLIILP AENGFDALRR QVPVRYSVRG GKVIASTQPA QTTVYLEQPE AIDYKR
Greater than 90% as determined by SDS-PAGE.
944996
50 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers