missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ssbF Protein
A DNA sequence encoding the Escherichia coli (strain K12) ssbF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7083-100ug
This item is not returnable.
View return policy
Specifications
NP_061439.1 | |
ssbF Protein | |
Research Use Only | |
ssbF | |
Recombinant Protein | |
E. coli | |
AVRGINKVIL VGRLGKDPEV RYIPNGGAVA NLQVATSESW RDKQTGEMRE QTEWHRVVLF GKLAEVAGEC LRKGAQVYIE GQLRTRSWED NGITRYVTEI LVKTTGTMQM LVRAAGAQTQ PEEGQQFSGQ PQPEPQAEAG TKKGGAKTKG RGRKAAQPEP QPQPPEGDDY GFSDDIPF | |
Greater than 90% as determined by SDS-PAGE. |
1263584 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |