missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr DUTPase Protein
A DNA sequence encoding the Epstein-Barr virus (strain GD1) (HHV-4) (Human herpesvirus 4) DUTPase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7034-1mg
This item is not returnable.
View return policy
Specifications
DUTPase Protein | |
Research Use Only | |
BLLF3 | |
Recombinant Protein | |
E. coli | |
MEACPHIRYA FQNDKLLLQQ ASVGRLTLVN KTTILLRPMK TTTVDLGLYA RPPEGHGLML WGSTSRPVTS HVGIIDPGYT GELRLILQNQ RRYNSTLRPS ELKIHLAAFR YATPQMEEDK GPINHPQYPG DVGLDVSLPK DLALFPHQTV SVTLTVPPPS IPHHRPTIFG RSGLAMQGIL VKPCRWRRGG VDVSLTNFSD QTVFLNKYRR FCQLVYLHKH HLTSFYSPHS DAGVLGPRSL FRWASCAFEE VPSLAMGDSG LSEALEGRQG RGFGSSGQ | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |