missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Acetylcholinesterase/ACHE Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 7574-CE-010

Product Code. 18168430

  • 591.00€ / 10µg
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Measured by its ability to cleave Acetylthiocholine. The specific activity is >500 nmol/min/μg, as measured under the described conditions.
Specifications

Specifications

P22303
Supplied as a 0.2 μm filtered solution in Tris and NaCl.
M.W. (Observed): 60-75 kDa, reducing conditions; M.W. (theoretical): 65 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
ACHE
Recombinant
Acetylcholinesterase/ACHE
Enzyme Activity
43
Acetylcholinesterase/ACHE
Chinese Hamster Ovary cell line, CHO-derived human Acetylcholinesterase/ACHE protein Glu32-Leu614, with a C-terminal 6-His tag
<0.10 EU / 1 μg of the protein by the LAL method.
Unconjugated
>90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.