Learn More
R&D Systems™ Recombinant Human Activin AC Heterodimer Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
386.00€
Specifications
Molecular Weight (g/mol) | 12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit) |
---|---|
Name | Activin AC |
Quantity | 10 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human Activin AC protein, (N-terminus) Human Activin beta A (Gly311-Ser426) Accession &Num; NP_002183 Human Activin beta C (Gly237-Ser352) Accession &Num; NP_005529 (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18309417
|
R&D Systems™
4879-AC-010 |
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
386.00€
10µg |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
18320328
|
R&D Systems™
4879-AC-010/CF |
Lyophilized from a 0.2μm filtered solution in HCl. |
386.00€
10µg |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
Description
The Recombinant Human Activin AC Heterodimer Protein is derived from CHO. The Recombinant Human Activin AC Heterodimer Protein has been validated for the following applications: Binding Activity.Specifications
12.9 kDa (betaA subunit), 12.5 kDa (betaC subunit) | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Activin AC | |
Chinese Hamster Ovary cell line,CHO-derived human Activin AC protein, (N-terminus) Human Activin beta A (Gly311-Ser426) Accession &Num; NP_002183 Human Activin beta C (Gly237-Ser352) Accession &Num; NP_005529 (C-terminus) | |
Disulfide-linked heterodimer | |
Recombinant |